Order Entry
Netherlands
ContactUsLinkComponent
NucleoBond 96 Xtra EF (4x96) 4x96 preps for the isolation of endotoxin free plasmid DNA-NucleoBond Filter Plate, NucleoBond Xtra EF Plate, NucleoBond Finalizer Plate, MN Wash Plate, Square-well Block, Culture Plate, Gas-permeable Foil, Elution Plate 1 * 384 items
Catalog # MANA740430.4
Supplier:  MACHEREY-NAGEL
NucleoBond 96 Xtra EF (4x96) 4x96 preps for the isolation of endotoxin free plasmid DNA-NucleoBond Filter Plate, NucleoBond Xtra EF Plate, NucleoBond Finalizer Plate, MN Wash Plate, Square-well Block, Culture Plate, Gas-permeable Foil, Elution Plate 1 * 384 items
Catalog # MANA740430.4
Supplier:  MACHEREY-NAGEL
Frequently Bought Together