Order Entry
Netherlands
ContactUsLinkComponent
Stericlin sterilisation tape, beige, with STEAM indicator (stripes), for sealing of crepe or non-woven or SMS wrapping materials, 1 9mm X 50
Catalog # VEPA3FKLB410102_U
Stericlin sterilisation tape, beige, with STEAM indicator (stripes), for sealing of crepe or non-woven or SMS wrapping materials, 1 9mm X 50
Catalog # VEPA3FKLB410102_U