Order Entry
Netherlands
ContactUsLinkComponent
Software conforming to FDA AtmoCONTROL. Meets the requirements for the use of electronically stored data sets and electronic signatu res as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence for the cont 1 * 1 items
Catalog # MEMMFDAQ1IF160PLUS
Supplier:  MEMMERT
Software conforming to FDA AtmoCONTROL. Meets the requirements for the use of electronically stored data sets and electronic signatu res as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence for the cont 1 * 1 items
Catalog # MEMMFDAQ1IF160PLUS
Supplier:  MEMMERT
Frequently Bought Together