Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
25 µL
Catalog # NOVUNBP1-42827SS
Supplier: Novus Biologicals
Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
Catalog # NOVUNBP1-42827SS
Supplier: Novus Biologicals
Jump to:
Product Details & Documents
Type: Primary
Antigen: MAP1B (microtubule-associated protein 1B)
Clonality: polyclonal
Clone: SuperBUGS
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Documents
NBP1-42827.pdf
Datasheet
Specifications
- Size:25 µL
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Microtubule-associated protein 1B
- Cat. No.:NOVUNBP1-42827SS
- Reactivity:Human
- Gene ID:4131
- Antigen synonyms:MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
- Conjugation:Unconjugated
- Antigen symbol:MAP1B
- Clone:SuperBUGS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



