Order Entry
Netherlands
ContactUsLinkComponent
IL17F Human Recombinant produced in E.coli is homodimeric, cysteine linked, non-glycosylated polypeptide chain containing x 134 amin o acids and having total molecular mass of 30.1 kDa.The IL-17F Human is purified by proprietary chromatograp 1 * 5 µG
Catalog # BIRBORB80078-5
Supplier:  Biorbyt
IL17F Human Recombinant produced in E.coli is homodimeric, cysteine linked, non-glycosylated polypeptide chain containing x 134 amin o acids and having total molecular mass of 30.1 kDa.The IL-17F Human is purified by proprietary chromatograp 1 * 5 µG
Catalog # BIRBORB80078-5
Supplier:  Biorbyt