Order Entry
Netherlands
ContactUsLinkComponent
IL3 Canine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 120 amino acids and having a mo lecular mass of 14 kDa. The IL-3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80060-10
Supplier:  Biorbyt
IL3 Canine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 120 amino acids and having a mo lecular mass of 14 kDa. The IL-3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80060-10
Supplier:  Biorbyt