Order Entry
Netherlands
ContactUsLinkComponent
Insulin-Like Growth Factor Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino a cids and having a molecular mass of 7687 Dalton. IGF-I is purified by proprietary chromatographic techniques 1 * 50 µG
Catalog # BIRBORB80047-50
Supplier:  Biorbyt
Insulin-Like Growth Factor Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino a cids and having a molecular mass of 7687 Dalton. IGF-I is purified by proprietary chromatographic techniques 1 * 50 µG
Catalog # BIRBORB80047-50
Supplier:  Biorbyt