Order Entry
Netherlands
ContactUsLinkComponent
The Recombinant Human S100P has molecular mass of 10.4 kDa containing 95 amino acid residues of the human S100P.MTELEAAMGM IIDVFSRYS G SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK. 1 * 10 µG
Catalog # BIRBORB81345-10
Supplier:  Biorbyt
The Recombinant Human S100P has molecular mass of 10.4 kDa containing 95 amino acid residues of the human S100P.MTELEAAMGM IIDVFSRYS G SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK. 1 * 10 µG
Catalog # BIRBORB81345-10
Supplier:  Biorbyt