Order Entry
Netherlands
ContactUsLinkComponent
GMFB Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 162 amino acids (1-142 a.a.)and having total molecular mass of 18.8 kDa. GMGB is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 20 µG
Catalog # BIRBORB80376-20
Supplier:  Biorbyt
GMFB Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 162 amino acids (1-142 a.a.)and having total molecular mass of 18.8 kDa. GMGB is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 20 µG
Catalog # BIRBORB80376-20
Supplier:  Biorbyt