Order Entry
Netherlands
ContactUsLinkComponent
MCSF Human Recombinant produced in E.coli is disulfide linked homodimer, glycosylated, polypeptide chain containing x 149 amino acid s and having total molecular mass of 42 kDa.MCSF is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80517-10
Supplier:  Biorbyt
MCSF Human Recombinant produced in E.coli is disulfide linked homodimer, glycosylated, polypeptide chain containing x 149 amino acid s and having total molecular mass of 42 kDa.MCSF is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80517-10
Supplier:  Biorbyt