Order Entry
Netherlands
ContactUsLinkComponent
Peri pump tube, 3 bridged grey/grey 6/pk. santoprene for use with sps 3 autosampler when pumping ketone-based solvent rinse. 1 * 6 items
Catalog # VARI3710057500
Supplier:  VARIAN
Peri pump tube, 3 bridged grey/grey 6/pk. santoprene for use with sps 3 autosampler when pumping ketone-based solvent rinse. 1 * 6 items
Catalog # VARI3710057500
Supplier:  VARIAN
Frequently Bought Together