Order Entry
Netherlands
ContactUsLinkComponent
SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: L DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 9,5 CM, DIAMETER OF TOP 21 CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M, CAP 36G/M, HAIRNET 10-12G/M PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CA 1 * 1.000 items
Catalog # GUNA130466
Supplier:  VWR Collection
SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: L DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 9,5 CM, DIAMETER OF TOP 21 CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M, CAP 36G/M, HAIRNET 10-12G/M PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CA 1 * 1.000 items
Catalog # GUNA130466
Supplier:  VWR Collection
Frequently Bought Together