Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Melatonin

Melatonin

Glass bottle for solids, 10 g
Catalog # CAYM14427-10
Supplier:   Cayman Chemical
Melatonin
Catalog # CAYM14427-10
Supplier:  Cayman Chemical
CAS Number:  73-31-4
Melatonin

Glass bottle for solids, 10 g

CAS Number:  73-31-4
Price
109,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • EINECS:200-797-7

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.