Phloretin
Glass bottle for solids, 250 mg
Catalog # CAYM14452-250
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:60-82-2
- Formula:C₁₅H₁₄O₅
- MDL Number:MFCD00002288
- Molecular Weight:274.28 g/mol
- EINECS:200-488-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



