Order Entry
Netherlands
ContactUsLinkComponent
Stand-alone Single wavelength slide-out transilluminator (20 x 20cm 302nm) 230V, 50Hz. Mediumwave (no cover) 6 tubes. For use with DG or on Benchtop as stand-alone. Includes mains cable 1 * 1 items
Catalog # SYNOBT-2020M-E
Stand-alone Single wavelength slide-out transilluminator (20 x 20cm 302nm) 230V, 50Hz. Mediumwave (no cover) 6 tubes. For use with DG or on Benchtop as stand-alone. Includes mains cable 1 * 1 items
Catalog # SYNOBT-2020M-E
Frequently Bought Together