Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, straight bore, 4,0 mm
Catalogus # BIBBSST104K
Leverancier: SciLabware
Ga naar:
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
- Supplied complete with plastic screw thread retaining device
- Self lubricating, avoiding the need for stopcock grease
- Made to a taper of 1:10 giving a combination of performance and ease of operation
Delivery information: Key is supplied complete with plastic screw thread retaining device.
Specificaties
- Hole Ø:4,0 mm
- Description:Spare PTFE keys for stopcocks, PYREX®, straight bore
- Seq:03
Aanbevelingen