Order Entry
Netherlands
ContactUsLinkComponent
6 components 10mg/l each of Ba Bi Ce In Li U in HNO3 2%, in aluminium bag. Shelf life: 6 months of opening the bottle, Length of validity: 12 months. 1 * 100 mL
Catalog # VWRS73DE.10.2N.L1
Supplier:  VWR Collection
6 components 10mg/l each of Ba Bi Ce In Li U in HNO3 2%, in aluminium bag. Shelf life: 6 months of opening the bottle, Length of validity: 12 months. 1 * 100 mL
Catalog # VWRS73DE.10.2N.L1
Supplier:  VWR Collection
Frequently Bought Together