Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Morpholine for synthesis, Sigma-Aldrich®

Morpholine for synthesis, Sigma-Aldrich®

Plastic bottle for solids, 10 kg
Catalog # 8.06127.9010
Flammable
Toxic
Corrosive
Supplier:   Merck
Morpholine for synthesis, Sigma-Aldrich®
Catalog # 8.06127.9010
Supplier:  Merck
CAS Number:  110-91-8
Morpholine for synthesis, Sigma-Aldrich®
Morpholine for synthesis, Sigma-Aldrich®

Plastic bottle for solids, 10 kg

CAS Number:  110-91-8
Price
Inquire For Price
Log in for availability
1 * 10 kg
Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:12,06362
  • Boiling Pt:128.9 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:1 g/cm³ (20 °C)
  • Flash Pt:38 °C
  • Storage Temperature:Ambient
  • UN:2054
  • ADR:8 (3),I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-815-1

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    87.12 g/mol
  • Boiling Point:
    128 - 130 °C (1013 hPa)
  • Vapor Pressure:
    10 hPa (20 °C)
  • Flash Point:
    32 °C
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    10.6 (5 g/l, H2O, 20 °C)
  • Ignition Point:
    275 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.