Order Entry
Netherlands
ContactUsLinkComponent
lysate, overexpression lysate, agpat2 293t cell transient overexpression lysate(denatured), 1 * 100 µl
Catalog # ABNOH00010555-T01
Supplier:  Abnova
lysate, overexpression lysate, agpat2 293t cell transient overexpression lysate(denatured), 1 * 100 µl
Catalog # ABNOH00010555-T01
Supplier:  Abnova