Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Test kits, Volatile organic acids, Spectroquant®
Test kits, Volatile organic acids, Spectroquant®
Volatile Organic acids cell test, 50 - 3000 mg/l CH₃COOH, Hydroxamic acids/ iron (III) salts, Prove, Nova 30/60, Multy, Move, 1
Catalogus # 1.01749.0001
Leverancier:   Merck
Test kits, Volatile organic acids, Spectroquant®
Catalogus # 1.01749.0001
Leverancier:  Merck
CAS-nummer:  
Test kits, Volatile organic acids, Spectroquant®
Test kits, Volatile organic acids, Spectroquant®
Volatile Organic acids cell test, 50 - 3000 mg/l CH₃COOH, Hydroxamic acids/ iron (III) salts, Prove, Nova 30/60, Multy, Move, 1
CAS-nummer:  
Price
166,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specificaties
Specificaties
  • Chemical method:
    Hydroxamic acids/ iron (III) salts
  • Pk:
    1
  • Description:
    Volatile Organic acids cell test
  • For unit:
    Prove, Nova 30/60, Multy, Move
  • Range:
    50 - 3000 mg/l CH₃COOH
Specificaties
Aanbevelingen