Order Entry
Netherlands
ContactUsLinkComponent
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.900-0.950, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F , with calibration certificate. 1 * 1 items
  ALLA3510LC09060BEU
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.900-0.950, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F , with calibration certificate. 1 * 1 items
  ALLA3510LC09060BEU
Frequently Bought Together