20-Hydroxyecdysone
10 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 5289-74-7 Molecular Formula: C27H44O7 Formula Weight: 480.64 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 242-244° Solubility: Soluble in alcohol Beilstein Reference: 1917578 MDL No.: MFCD00036740
Applications: An insect steroid hormone that induces apoptosis
Synonyms: β-Ecdysone
- CAS Number:5289-74-7
- Formula:C₂₇H₄₄O₇
- MDL Number:MFCD00036740
- Molecular Weight:480.642 g/mol
- Merck Index:14,3491
- Melting Pt:242…244 °C
Documents
Specifications
Specifications
- Pk:10 mg



