Order Entry
Netherlands
Orders LinkContactUsLinkComponent
CF® Dye cAMP

CF® Dye cAMP

CF® 488A cAMP, 490/515 nm, 100 µG
Catalog # 00036.
Supplier:   Biotium
CF® Dye cAMP
Catalog # 00036.
Supplier:  Biotium
CAS Number:  
CF® Dye cAMP

CF® 488A cAMP, 490/515 nm, 100 µG

CAS Number:  
Price
204,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

Fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.

  • Superior CF® dyes are bright, photostable and water-soluble
  • Available with green or far-red labels
  • Soluble in H₂O, DMF, or DMSO

CF®488A-cAMP and CF®640R-cAMP are fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.

Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.

CF is a registered trademark of Biotium, Inc. Alexa Fluor, Texas Red, and DyLight are registered trademarks of Thermo Fisher Scientific.

Specifications

Specifications

  • Pk:
    100 µG
  • Description:
    CF® 488A cAMP
  • Ex/Em:
    490/515 nm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.