Order Entry
Netherlands
ContactUsLinkComponent
Extension collars for CoolBox XT Green. Place between the base and original collar to extend the height of the ice-free workstation and enable the use of taller CoolRack sample tube modules. Green. 1 * 1 items
Catalog # 479-1300
Supplier:  BIOCISION
Extension collars for CoolBox XT Green. Place between the base and original collar to extend the height of the ice-free workstation and enable the use of taller CoolRack sample tube modules. Green. 1 * 1 items
Catalog # 479-1300
Supplier:  BIOCISION
Frequently Bought Together