Order Entry
Netherlands
ContactUsLinkComponent
Agilent J&W PoraPLOT Q, 25 m x 0.53 mm id, 20 µm, GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWLCP7554PT
Supplier:  Agilent
Agilent J&W PoraPLOT Q, 25 m x 0.53 mm id, 20 µm, GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWLCP7554PT
Supplier:  Agilent
Frequently Bought Together