Polarising microscopes, BA310 POL
CCIS® plan achromatic objective (tension-free) EC PL P (WD = 0,35 mm)
Catalogus # MOTI1101001703981
Leverancier: MOTIC
Ga naar:
Product Details & Documents
These models offer higher optical performance and modularity.
- Polariser mounted under the condenser holder rotates 360° with click stop at 90°
- Highly stable, robust symmetrical pyramid design and T-shaped base
- Anti-fungal chemical treatment to inhibit germs growth
- Plan achromatic, stain-free objectives
- One eyepiece features a crosshair reticle
Suitable for life science, geology, material science and forensics.
Delivery information: Supplied with an Allen key, a blue filter and a dust cover.
Documents
BA310 POL Bino SP_1210_EN
Datasheet
BA310 POL_CA_1604_EN
Product Literature
Specificaties
- Magnification:60×/0,80/S
- Description:CCIS® plan achromatic objective (tension-free) EC PL P (WD = 0,35 mm)
- Seq:105
Accessories
Productinformatie
- Catalogus #DescriptionVoltagePowerBeschikbaarheidPrice
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
- 630-19450,65×C-mount camera adapter for ²/₃" chip sensorsLog in voor beschikbaarheid1 * 1 ST - 159,00 €
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
- 630-15030,5×C-mount camera adapter for ¹/₃ and ½" chip sensorsLog in voor beschikbaarheid1 * 1 ST - 159,00 €
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



