Order Entry
Netherlands
ContactUsLinkComponent
HSP 60 (SPM253) mouse monoclonal IgG 200 µg/ml raised against recombinant HSP 60 of human origin, with epitope mapping to amino a cids 383-447 1 * 200 µG
Catalog # SANTSC-65568
HSP 60 (SPM253) mouse monoclonal IgG 200 µg/ml raised against recombinant HSP 60 of human origin, with epitope mapping to amino a cids 383-447 1 * 200 µG
Catalog # SANTSC-65568
Frequently Bought Together