PMS2 (349.29.5.2) mouse monoclonal IgG1 100 µg/ml raised against recombinant fragment PMS2 corresponding to the first 133 amino a cid residues of human origin 1 * 100 µG
Catalog # SANTSC-56203
Supplier: Santa Cruz Biotechnology
Frequently Bought Together