Order Entry
Netherlands
ContactUsLinkComponent
Respiratory Syncytial Virus NP (B023) mouse monoclonal IgG1 100 µg/ml raised against Respiratory Syncytial Virus strains 127, SNK and 9007 of cow origin and Respiratory Syncytial Virus strains A2, Long, Randall and 8/60 of human origin 1 * 100 µG
  SANTSC-58001
Respiratory Syncytial Virus NP (B023) mouse monoclonal IgG1 100 µg/ml raised against Respiratory Syncytial Virus strains 127, SNK and 9007 of cow origin and Respiratory Syncytial Virus strains A2, Long, Randall and 8/60 of human origin 1 * 100 µG
  SANTSC-58001
Frequently Bought Together