Respiratory Syncytial Virus fusion protein (RSV3216) mouse monoclonal IgG2b 100 µg/ml raised against Respiratory Syncytial Virus strains 127, SNK and 9007 of cow origin and Respiratory Syncytial Virus strains A2, Long, Randall and 8/60 of human or 1 * 100 µG
Catalog # SANTSC-58000
Supplier: Santa Cruz Biotechnology
Frequently Bought Together