p-NHE-3 (10A8) mouse monoclonal IgG2b 200 µg/ml raised against a serine-phosphorylated synthetic peptide corresponding to amino a cids 594-615 of NHE-3 of rat origin 1 * 200 µG
Catalog # SANTSC-53961
Supplier: Santa Cruz Biotechnology
Frequently Bought Together