CagA (HP-317) mouse monoclonal IgG 100 µg/ml raised against a recombinant fragment (consisting of 580 C-terminal amino acid resid ues) of CA-gene of H.pylori origin 1 * 100 µG
Catalog # SANTSC-51791
Supplier: Santa Cruz Biotechnology
Frequently Bought Together