Amphiphysin II (D-3) mouse monoclonal IgG2a 200 µg/ml raised against amino acids 421-520 mapping within an internal region of Amp hiphysin II of human origin 1 * 200 µG
Catalog # SANTSC-74486
Supplier: Santa Cruz Biotechnology
Frequently Bought Together