Order Entry
Netherlands
ContactUsLinkComponent
MIP-4 (H-12) mouse monoclonal IgG1 200 µg/ml specific for an epitope mapping between amino acids 61-89 at the C-terminus of MIP-4 of human origin 1 * 200 µG
  SANTSC-374439
MIP-4 (H-12) mouse monoclonal IgG1 200 µg/ml specific for an epitope mapping between amino acids 61-89 at the C-terminus of MIP-4 of human origin 1 * 200 µG
  SANTSC-374439
Frequently Bought Together