p-PACT (1A6-1H4) mouse monoclonal IgG1 200 µg/ml raised against a synthetic peptide corresponding to amino acids 13 to 25, includ ing phosphorylated serine 18, of mouse PACT protein 1 * 200 µG
Catalog # SANTSC-73478
Supplier: Santa Cruz Biotechnology
Frequently Bought Together