17beta-HSD2 (H-12) mouse monoclonal IgG2a 200 µg/ml raised against amino acids 266-375 mapping near the C-terminus of 17&beta -HS D2 of human origin 1 * 200 µG
Catalog # SANTSC-373990
Supplier: Santa Cruz Biotechnology
Frequently Bought Together