Order Entry
Netherlands
ContactUsLinkComponent
17-beta Estradiol (5E500) mouse monoclonal IgG1 100 µg/ml raised against 17 beta Estradiol conjugated to BSA 1 * 100 µG
Catalog # SANTSC-69961
17-beta Estradiol (5E500) mouse monoclonal IgG1 100 µg/ml raised against 17 beta Estradiol conjugated to BSA 1 * 100 µG
Catalog # SANTSC-69961
Frequently Bought Together