Order Entry
Netherlands
ContactUsLinkComponent
Troponin I-C (415) mouse monoclonal IgG1 100 µg/ml raised against cardiac troponin I of human origin with epitope mapping to amin o acids 122-139 1 * 100 µG
Catalog # SANTSC-66123
Troponin I-C (415) mouse monoclonal IgG1 100 µg/ml raised against cardiac troponin I of human origin with epitope mapping to amin o acids 122-139 1 * 100 µG
Catalog # SANTSC-66123
Frequently Bought Together