Morpholine ≥99.0% ACS
4
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.0% min CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972
- CAS Number:110-91-8
- Formula:C₄H₉NO
- MDL Number:MFCD00005972
- Molecular Weight:87.12 g/mol
- Merck Index:14,6277
- Boiling Pt:129 °C
- Melting Pt:–5 °C
- Density:1.007
- Flash Pt:35 °C (95 °F)
- Storage Temperature:Ambient
- EINECS:203-815-1
- UN:2054
- ADR:8,I
Documents
Specifications
Specifications
- Pk:4



