Order Entry
Netherlands
ContactUsLinkComponent
28 COMPOSES A 100 MG/L CHAQUE DANS HNO3 5% Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn
Catalog # 89143.260
Supplier:  BDH Prolabo
28 COMPOSES A 100 MG/L CHAQUE DANS HNO3 5% Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn
Catalog # 89143.260
Supplier:  BDH Prolabo
Frequently Bought Together