Order Entry
Netherlands
ContactUsLinkComponent
olumetric flasks with one circular mark, 10 ml, +/- 0,04 ml, class A ,borosilicate glass 3.3, clear glass, blue graduated, with ground jointand polystopper, NS 10/19, 10 pcs./carton, MARIENFELD SUPERIOR GTIN: 4250317305695 Customs tariff no.: 7017200 1 * 10 items
Catalog # MARI2332191
Supplier:  MARIENFELD
olumetric flasks with one circular mark, 10 ml, +/- 0,04 ml, class A ,borosilicate glass 3.3, clear glass, blue graduated, with ground jointand polystopper, NS 10/19, 10 pcs./carton, MARIENFELD SUPERIOR GTIN: 4250317305695 Customs tariff no.: 7017200 1 * 10 items
Catalog # MARI2332191
Supplier:  MARIENFELD
Frequently Bought Together