Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, FGFR4 (HUMAN) RECOMBINANT PROTEIN, HUMAN FGFR4 (NM_002011, 391 A.A. - 802 A.A.) PARTIAL RECOMBINANT PROT EIN WITH GST-HIS TAG EXPRESSED IN SF9 CELLS. 1 * 100 µG
ABNOP4693
:
Abnova
RECOMBINANT/NATIVE PROTEIN, FGFR4 (HUMAN) RECOMBINANT PROTEIN, HUMAN FGFR4 (NM_002011, 391 A.A. - 802 A.A.) PARTIAL RECOMBINANT PROT EIN WITH GST-HIS TAG EXPRESSED IN SF9 CELLS. 1 * 100 µG
ABNOP4693
:
Abnova
TDS_ABNOP4693.pdf
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals