Order Entry
Netherlands
ContactUsLinkComponent
KIT, CYCLIC AMP COMPLETE ELISA KIT, CYCLIC AMP COMPLETE ELISA KIT IS A KIT FOR THE QUANTITATIVE DETERMINATION OF CYCLIC AMP IN CELLS AND TISSUE TREATED WITH 0.1M HCL, IN ADDITION TO CULTURE SUPERNATES, SALIVA, AND SERUM. 1 * 1 KIT
Catalog # ABNOKA0320
Supplier:  Abnova
KIT, CYCLIC AMP COMPLETE ELISA KIT, CYCLIC AMP COMPLETE ELISA KIT IS A KIT FOR THE QUANTITATIVE DETERMINATION OF CYCLIC AMP IN CELLS AND TISSUE TREATED WITH 0.1M HCL, IN ADDITION TO CULTURE SUPERNATES, SALIVA, AND SERUM. 1 * 1 KIT
Catalog # ABNOKA0320
Supplier:  Abnova