Order Entry
Netherlands
ContactUsLinkComponent
KIT, RED PHOSPHATASE SUBSTRATE KIT, RED PHOSPHATASE SUBSTRATE KIT CONTAINS REAGENTS FOR THE DETECTION OF ALKALINE PHOSPHATASE-LABELE D CONJUGATES. RED REAGENTS FORM A BRILLIANT SCARLET REACTION PRODUCT THAT IS STABLE IN ORGANIC SOLVENTS. SECTIONS REA 1 * 1 KIT
Catalog # ABNOKA0566
Supplier:  Abnova
KIT, RED PHOSPHATASE SUBSTRATE KIT, RED PHOSPHATASE SUBSTRATE KIT CONTAINS REAGENTS FOR THE DETECTION OF ALKALINE PHOSPHATASE-LABELE D CONJUGATES. RED REAGENTS FORM A BRILLIANT SCARLET REACTION PRODUCT THAT IS STABLE IN ORGANIC SOLVENTS. SECTIONS REA 1 * 1 KIT
Catalog # ABNOKA0566
Supplier:  Abnova
Frequently Bought Together