Order Entry
Netherlands
ContactUsLinkComponent
KIT, S100B (HUMAN) ELISA KIT, S100B (HUMAN) ELISA KIT IS A HRP LABELLED ANTIBODY BASED SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITAT IVE MEASUREMENT OF HUMAN S100B PROTEIN JUST ONLY FOR SERUM SAMPLES, HEPARIN PLASMA SAMPLES AND CEREBROSPINAL FLUID SA 1 * 1 KIT
Catalog # ABNOKA0037
Supplier:  Abnova
KIT, S100B (HUMAN) ELISA KIT, S100B (HUMAN) ELISA KIT IS A HRP LABELLED ANTIBODY BASED SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITAT IVE MEASUREMENT OF HUMAN S100B PROTEIN JUST ONLY FOR SERUM SAMPLES, HEPARIN PLASMA SAMPLES AND CEREBROSPINAL FLUID SA 1 * 1 KIT
Catalog # ABNOKA0037
Supplier:  Abnova
Frequently Bought Together