Order Entry
Netherlands
ContactUsLinkComponent
Glass tube for LRC15 column (15mm id) 80-200mm for Pall Laboratory LRC empty chromatography column - Pall Part Number LRC15XTUBE0 80 -200 1 * 1 items
Catalog # 554-2376
Glass tube for LRC15 column (15mm id) 80-200mm for Pall Laboratory LRC empty chromatography column - Pall Part Number LRC15XTUBE0 80 -200 1 * 1 items
Catalog # 554-2376
Frequently Bought Together