Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Dexamethasone

Glass bottle for solids, 5 g
Catalog # CAYM11015-5
Supplier:   Cayman Chemical
Dexamethasone
Catalog # CAYM11015-5
Supplier:  Cayman Chemical
CAS Number:  50-02-2
Dexamethasone

Glass bottle for solids, 5 g

CAS Number:  50-02-2
Price
617,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Formula:C₂₂H₂₉FO₅
  • MDL Number:MFCD00064136
  • Molecular Weight:392.47 g/mol
  • Melting Pt:268…271 °C
  • EINECS:200-003-9

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications