Dexamethasone
Glass bottle for solids, 500 mg
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
- CAS Number:50-02-2
- Formula:C₂₂H₂₉FO₅
- MDL Number:MFCD00064136
- Molecular Weight:392.47 g/mol
- Melting Pt:268…271 °C
- EINECS:200-003-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



