Capillary columns, Chromolith® CapRod®
Capillary, RP-8e Trap, 0,10 mm, 50 mm
: Merck
Fast nano-LC for proteins and peptides. Chromolith® CapRod® capillaries combine the speed of monolithic silica technology with the sensitivity of nano-LC, thus enabling high speed high sensitivity LC-MS analytics.
- High flow rates at low pressure
- Long column lifetime
- Robust handling
- Column Format:Capillary
- Ø int.:0,10 mm
- Length:50 mm
- Seq:09
- Phase:RP-8e Trap



