Water purification system, AFS® 10E/15E
AFS® 10E water purification system (EU plug, manual in Italian)
Catalog # MILLZAFS10E0IT
Jump to:
Discontinued
Product Details & Documents
Economical water system with Elix® technology, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 L/min. For clinical analysers with pure water needs up to 300 L daily.
- Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality
- Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
- No resin cylinders to replace: Lower running costs
- Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
- Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
Patented Elix® technology provides constant quality water without the need for softeners or conditioning cartridges, which means reduced maintenance― and less analyser downtime.
Specifications
- Description:AFS® 10E water purification system (EU plug, manual in Italian)
- Seq:06
- Cat. No.:MILLZAFS10E0IT
- Production flow rate:10 L/h @ 25 °C
- Feed water:Portable tap water at a maximum pressure of 6 bar
- Model:AFE® 10E
- Bacteria:< 1 CFU/ml
- W×D×H:268×426×585 mm
- Resistivity at +25 °C:>15 MΏ.cm
- TOC content:< 30 ppb
- Weight:28 kg



